Learn More
Carbosynth LLC. Fmoc-mini-PEGTM Fmoc-8-Amino-3,6-Dioxaoctanoic Acid, 5g, MW: (M.W. 385.42), Molecular formula: C21H23NO6, CAS# [166108-71-0]
SDP
Supplier:  Carbosynth LLC. FXX5521PI5G
Encompass_Preferred
Synonyms: Fmoc-8-Amino-3,6-Dioxaoctanoic Acid 9-Fluorenylmethoxycarbonyl-8-Amino-3,6-Dioxaoctanoic Acid, Fmoc-AEEA, Storage Temperature: 2-10°C
Catalog No. 50-168-5778