Learn More
Medchemexpress LLC Fmoc-MMAF-OMe | 863971-38-4 | 98.6% | C55H77N5O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY7925550MG
Encompass_Preferred
Fmoc-MMAF-OMe is an anticancer agent and tubulin polymerization inhibitor, featuring an Fmoc protecting group. Its active ingredient, MMAF, is a cytotoxic component commonly found in classic antibody drug conjugates.
  • Anticancer agent
  • Tubulin polymerization inhibitor
  • Features an Fmoc protecting group
  • Active ingredient MMAF is a cytotoxic component of ADCs
  • For research use only
Catalog No. 50-004-33008