Learn More
Medchemexpress LLC Fmoc-N-(2-Boc-aminoethyl)-Gly-OH | 141743-15-9 | MFCD02259490 | 99.8% | 440.49 g·mol⁻1 | C24H28N2O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW101718250MG
Encompass_Preferred
Fmoc-N-(2-Boc-aminoethyl)-Gly-OH is an Fmoc-protected glycine derivative used as a building block in antibody-drug conjugate (ADC) linker synthesis and peptide coupling. It provides an Fmoc-protected amino terminus and a Boc-protected aminoethyl side chain to allow selective deprotection and coupling steps in medicinal chemistry and linker assembly.
  • Fmoc-protected amino terminus for standard peptide coupling.
  • Boc-protected aminoethyl side chain enabling selective deprotection.
  • High purity for reliable synthesis.
  • Suitable for ADC linker and peptide synthesis applications.
  • Available in common laboratory quantities, including 250 mg.
Catalog No. 50-002-04009