Learn More
eMolecules​ Fmoc-N-amido-PEG6-acid | Broadpharm | 882847-34-9 | MFCD06656474 | 575.655 | C30H41NO10 | 98.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 100mg | 208939966
Supplier:  eMolecules​ 208939966
Encompass_Preferred
Fmoc-N-amido-PEG6-acid | Broadpharm | 882847-34-9 | MFCD06656474 | 575.655 | C30H41NO10 | 98.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 100mg | 208939966
Catalog No. NC2948381