Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-N-Me-Ile-OH | 138775-22-1 | 99.9% | 367.44 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0085555G

Encompass_Preferred

Fmoc-N-Me-Ile-OH is an amino acid derivative primarily used in peptide synthesis.

  • Appears as a white to off-white solid
  • Molecular weight: 367.44
  • Chemical formula: C22H25NO4
  • Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 25 mg/mL

Catalog No. 50-004-23464


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.