Learn More
Medchemexpress LLC Fmoc-N-Me-Phe-OH | 77128-73-5 | MFCD00151938 | 99.8% | 401.46 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01098610MM/1ML
Encompass_Preferred
Fmoc-N-Me-Phe-OH is an N-methylated amino acid that can be used for the synthesis of some active compounds. It can be used in peptide coupling reactions.
  • N-methylated amino acid
  • Can be used for the synthesis of some active compounds
  • Can be used in peptide coupling reactions
  • For research use only
Catalog No. 50-004-53861