Learn More
Medchemexpress LLC Fmoc-N(Me)-Sar10 | 2375600-56-7 | 98.2% | 951.03 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP4198
Encompass_Preferred
Fmoc-N(Me)-Sar10 is an Fmoc-protected derivative of a methylated sarcosine decamer, primarily used in peptide synthesis to introduce enzymatically stable spacer sequences. It supports cell adhesion, proliferation, and maintenance of cell phenotype by mimicking the extracellular matrix (ECM), making it suitable for 3D cell culture and tissue engineering applications, especially with chondrocytes.
  • Used in peptide synthesis
  • Introduces enzymatically stable spacer sequences
  • Supports cell adhesion, proliferation, and phenotype maintenance
  • Mimics extracellular matrix for 3D growth
  • Suitable for tissue engineering and 3D cell culture
  • Functions as a building block for peptide ligand-conjugates
  • Enhances enzymatic stability of target peptides
  • Acts as a spacer for bioactive peptides and functional reagents
Catalog No. 50-003-98523