Learn More
Medchemexpress LLC Fmoc-N(Me)-Sar10 | 2375600-56-7 | 98.2% | 951.03 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP419850MG
Encompass_Preferred
Fmoc-N(Me)-Sar10 is an Fmoc-protected derivative of a methylated sarcosine decamer. It mimics the extracellular matrix (ECM) to provide a 3D growth microenvironment for cells.
  • Supports cell adhesion, proliferation, and maintenance of cell phenotype.
  • Primarily used in peptide synthesis to introduce enzymatically stable spacer sequences.
  • Suitable for tissue engineering and 3D cell culture.
  • Particularly suitable for in vitro culture studies of cells such as chondrocytes.
Catalog No. 50-003-97631