Learn More
Medchemexpress LLC Fmoc-N-me-Trp(Boc)-OH | 197632-75-0 | 540.61 | 5 G
Supplier: Medchemexpress LLC HYW0070565G
Fmoc-N-me-Trp(Boc)-OH is an amino acid derivative with an Fmoc protecting group. It can be used to synthesize peptide antibiotics with antibacterial activity against Pseudomonas aeruginosa.
- Amino acid derivative with Fmoc protecting group
- Used to synthesize peptide antibiotics
- Active against Pseudomonas aeruginosa
- For research use only