Learn More
Medchemexpress LLC Fmoc-N-PEG20-acid | 1952360-93-8 | C58H97NO24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140462
Encompass_Preferred
Fmoc-N-amido-PEG20-acid is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52512