Learn More
Medchemexpress LLC Fmoc-N-PEG20-acid | 1952360-93-8 | C58H97NO24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14046250MG
Encompass_Preferred
Fmoc-N-amido-PEG20-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53064