Learn More
Medchemexpress LLC Fmoc-N-PEG24-acid | 2170484-59-8 | 99.9% | 5 G

Supplier: Medchemexpress LLC HY1309075G
Fmoc-N-PEG24-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound is for research use only.
- Can be used in the synthesis of PROTACs
- High purity of 99.9%
- Molecular weight: 1368.59
- Appearance: solid
- Color: white to off-white
- Soluble in DMSO at 100 mg/mL
- Store at 4°C, sealed, away from moisture
- Related to cancer and cancer targeted therapy research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.