Learn More
Medchemexpress LLC Fmoc-NH-PEG1-C2-acid | 1654740-73-4 | 99.8% | 355.38 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140013
Encompass_Preferred
Fmoc-NH-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
Catalog No. 50-003-47836