Learn More
Medchemexpress LLC Fmoc-NH-PEG11-CH2CH2COOH | 3076038-46-2 | 98.5% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130870100MG
Encompass_Preferred
Fmoc-NH-PEG11-CH2CH2COOH is a PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs, which are molecules designed to degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-39806