Learn More
Medchemexpress LLC Fmoc-NH-PEG12-CH2CH2COOH | 1952360-91-6 | 100.0% | 839.97 | 5 G
SDP
Supplier:  Medchemexpress LLC 5GHY135821
Encompass_Preferred
Fmoc-NH-PEG12-CH2CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules containing two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. PROTACs work by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and complies with specifications.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-64442