Learn More
Medchemexpress LLC Fmoc-NH-PEG2-CH2CH2COOH | 872679-70-4 | 99.8% | 399.44 | C22H25NO6 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW04023825G
Encompass_Preferred
Fmoc-NH-PEG2-CH2CH2COOH is an Fmoc-protected PEG2-based linker that provides an Fmoc-protected amine and a terminal carboxylic acid functional group for use in bioconjugation and linker synthesis.
  • Provides Fmoc-protected amine and terminal carboxylic acid for conjugation
  • Peg2 spacer increases solubility and flexibility in linkers
  • Suitable for ADC and PROTAC linker synthesis and other bioconjugation applications
  • High purity (99.8%) suitable for research use
  • Available in multiple pack sizes, including 25 G
Catalog No. 50-002-03213