Learn More
Medchemexpress LLC HY-131955 250mg Medchemexpress, Fmoc-NH-PEG2-NH2 CAS:190249-87-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131955250MG
Encompass_Preferred
Medchemexpress, HY-131955 250mg Fmoc-NH-PEG2-NH2 CAS:190249-87-7 Fmoc-NH-PEG2-NH2 is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1300