Learn More
STA PHARMACEUTICAL US LLC Fmoc-NH-PEG24-CH2CH2COOH | 5 g | CAS 2170484-59-8 | MDL MFCD11041159
Supplier: STA PHARMACEUTICAL US LLC STAAA104095GM
Fmoc-NH-PEG24-CH2CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 2170484-59-8
- MDL: MFCD11041159
- InChIKey: NFOMZHGRFWXDGH-UHFFFAOYSA-N
- Molecular Weight: 1368.609
- Molecular Formula: C66H113NO28
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31,34,37,40,43,46,49,52,55,58,61,64,67,70,73,76-pentacosaoxa-4-azanonaheptacontan-79-oic acid
- SMILES: OC(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.