Learn More
Medchemexpress LLC Fmoc-NH-PEG4-alcohol | 868594-41-6 | 99.2% | 415.48 | 5 G
SDP
Supplier:  Medchemexpress LLC 5GHY133470
Encompass_Preferred
Fmoc-NH-PEG4-alcohol is a PEG-based PROTAC linker that facilitates the synthesis of PROTACs. These PROTACs are designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets a specific protein of interest. This mechanism enables PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-66248