Learn More
Medchemexpress LLC HY-W000434 100mg Medchemexpress, Fmoc-NH-PEG4-CH2CH2COOH CAS:557756-85-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW000434100MG
Encompass_Preferred
Medchemexpress, HY-W000434 100mg Fmoc-NH-PEG4-CH2CH2COOH CAS:557756-85-1 Fmoc-NH-PEG4-CH2CH2COOH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3198