Learn More
Medchemexpress LLC Fmoc-NH-PEG4-CH2COOH | 437655-95-3 | 99.1% | 473.52 | C25H31NO8 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1301755G
Encompass_Preferred
Fmoc-NH-PEG4-CH2COOH is an Fmoc-protected PEG4 linker bearing a terminal carboxylic acid. It is used as a cleavable ADC linker and as a PEG-based PROTAC linker for conjugation and linker synthesis. The compound has molecular formula C25H31NO8 and molecular weight 473.52 and is supplied at high purity for research use.
  • Fmoc-protected amine compatible with solid-phase and solution-phase synthesis.
  • Terminal carboxylic acid enables standard coupling reactions.
  • Peg4 spacer increases solubility and provides linker flexibility.
  • High purity suitable for research and conjugation chemistry.
  • Available in multiple sizes, including a 5 g package.
Catalog No. 50-002-03944