Learn More
Medchemexpress LLC Fmoc-NH-PEG9-CH2CH2COOH | 1191064-81-9 | 98.21% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1301675G
Encompass_Preferred
Fmoc-NH-PEG9-CH2CH2COOH is a cleavable linker utilized in the synthesis of antibody-drug conjugates (ADCs) and also serves as a PEG-based linker for the synthesis of PROTACs.
  • Cleavable linker
  • Used in antibody-drug conjugate (ADC) synthesis
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
Catalog No. 50-003-38954