Learn More
Medchemexpress LLC Fmoc-NMe-Val-Val-Dil-Dap-OH | 1352202-13-1 | MFCD00268593 | 96.2% | 807.03 g·mol⁻¹ | C45H66N4O9 | 100 MG

Supplier: Medchemexpress LLC HYW250408100MG
Fmoc-NMe-Val-Val-Dil-Dap-OH is a synthetic peptide intermediate used to prepare the MC-MMAF drug-linker conjugate for antibody-drug conjugate (ADC) synthesis. Supplied as a light-yellow solid for research use, the product carries documented purity, molecular weight, formula, and recommended storage conditions.
- Purity: 96.2%.
- Molecular weight: 807.03 g·mol⁻¹.
- Chemical formula: C45H66N4O9.
- Appearance: light yellow to yellow solid.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Intended use: intermediate for ADC drug-linker synthesis (research use only).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.