Learn More
Medchemexpress LLC Fmoc-O-(benzylphospho)-L-serine | 158171-14-3 | MFCD00797869 | 99.8% | 497.43 g/mol | C25H24NO8P | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017617
Encompass_Preferred
Fmoc-Ser(HPO3Bzl)-OH is an Fmoc-protected serine derivative bearing a benzyl-protected phosphate on the side chain. It is provided as a solid, high-purity building block for the incorporation of phosphorylated serine residues in synthetic peptides using standard Fmoc solid-phase peptide synthesis methods.
  • Fmoc-protected building block compatible with standard Fmoc SPPS.
  • Benzyl-protected phosphate preserves phosphorylation during synthesis.
  • High purity suitable for analytical and preparative peptide work.
  • Solid form enables convenient handling and storage.
  • Intended for incorporation of phospho-serine residues in peptides.
Catalog No. 50-003-02710