Learn More
Medchemexpress LLC Fmoc-O-methyl-D-tyrosine | 201335-88-8 | 99.6% | 417.45 g/mol | C25H23NO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02222710G
Encompass_Preferred
Fmoc-D-Tyr(Me)-OH is an Fmoc-protected O-methyl derivative of D-tyrosine used as a building block in solid-phase peptide synthesis. It provides side-chain protection to prevent undesired reactions during peptide assembly and is supplied for research use.
  • Fmoc-protected amino acid building block.
  • O-methylated tyrosine side chain for protection of the phenolic hydroxyl.
  • Suitable for solid-phase peptide synthesis and amino-acid modification.
  • High purity for research use (99.6%).
  • Available as solid and as ready-to-use DMSO solutions.
  • Stable when stored as powder at -20°C or 4°C; solutions stored at -80°C.
Catalog No. 50-003-00119