Learn More
Apexbio Technology LLC Fmoc-Osu 82911-69-1 100g
SDP
Supplier:  Apexbio Technology LLC A7012100000
Encompass_Preferred
Fmoc-Osu (82911-69-1) is a reagent widely used in peptide synthesis and biomolecular conjugation It introduces the reversible fluorenylmethoxycarbonyl (Fmoc) protective group onto primary amines by reacting via its activated N-hydroxysuccinimide ester temporarily masking amine functionalities This reversible protection enables controlled peptide elongation and sequential deprotection steps in solid-phase peptide synthesis Fmoc-Osu also facilitates selective modification of proteins peptides or biointerfaces in biomolecular conjugation strategies Based on these properties Fmoc-Osu supports research in peptide synthesis protein labeling and biological surface functionalization for biomedical applications
Catalog No. 50-000-94057