Learn More
TARGETMOL CHEMICALS INC FMOC-PEA 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1797710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Fmoc-PEA (Example 1-2) is a used as a cleavable linker for antibody-drug conjugates (ADC). purity: 99%
Catalog No. 50-258-1092