Learn More
Medchemexpress LLC Fmoc-PEG12-NHS ester | 2227246-92-4 | ≥98.0% | C46H68N2O18 | 25 MG

Supplier: Medchemexpress LLC 25MGHY141101
Fmoc-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. This design allows PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
- Appearance: Colorless to light yellow liquid
- Purity: ≥98.0% (NMR)
- Molecular weight: 937.03
- Recommended storage conditions for pure form: -20°C for 3 years
- Recommended storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
- Identified for laboratory use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.