Learn More
Medchemexpress LLC Fmoc-PEG12-NHS ester | 2227246-92-4 | 98.0% | C46H68N2O18 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141101
Encompass_Preferred
Fmoc-PEG12-NHS ester is a PEG-based PROTAC linker that plays a crucial role in the synthesis of PROTACs. This product is intended for research use only.
  • Acts as a PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-52589