Learn More
Medchemexpress LLC Fmoc-PEG12-NHS ester | 2227246-92-4 | ≥98.0% | C46H68N2O18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14110150MG
Encompass_Preferred
Fmoc-PEG12-NHS ester is a PEG-based PROTAC linker for research use only. It can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55142