Learn More
Medchemexpress LLC Fmoc-PEG2-C2-NHS ester | 1807534-85-5 | 96.08% | 100 MG

Supplier: Medchemexpress LLC HY126881100MG
Fmoc-PEG2-C2-NHS ester, with a molecular formula of C26H28N2O8 and molecular weight of 496.51, is a PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs (Proteolysis Targeting Chimeric Molecules). PROTACs are designed with two distinct ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins. This product is intended for research and analytical applications.
- PROTAC linker for targeted protein degradation
- Utilizes the ubiquitin-proteasome system
- Store pure form at -20°C for 3 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.