Learn More
Medchemexpress LLC Fmoc-PEG3-alcohol | 560088-66-6 | 98.0% | 371.43 g/mol | C21H25NO5 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW143484250MG
Encompass_Preferred
Fmoc-PEG3-alcohol is an Fmoc-protected PEG3 spacer bearing a terminal alcohol, provided as a solid building block for synthesis of PROTACs and other bifunctional molecules. It offers a short, flexible linker with an orthogonally protected handle to facilitate stepwise coupling and deprotection strategies.
  • Short PEG3 spacer for flexible linker geometry.
  • Terminal primary alcohol for further functionalization.
  • Fmoc-protected handle compatible with orthogonal deprotection.
  • Suitable for building PROTACs and other bifunctional molecules.
  • Available in milligram-to-gram pack sizes for synthesis scale-up.
  • High purity (reported 98%) suitable for research use.
Catalog No. 50-002-83809