Learn More
Medchemexpress LLC Fmoc-PEG3-alcohol | 560088-66-6 | MFCD08669697 | ≥95.0% | 371.43 g·mol⁻¹ | C21H25NO5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1434841G
Encompass_Preferred
Fmoc-PEG3-alcohol is an Fmoc-protected PEG3 alcohol used as a short, flexible linker building block for the synthesis of proteolysis-targeting chimeras (PROTACs) and other bifunctional molecules. It provides an Fmoc-protected amine handle compatible with common peptide and organic synthesis workflows.
  • Short, flexible PEG3 spacer suitable for linker design.
  • Fmoc-protected amine terminus for standard coupling chemistry.
  • High purity suitable for research and development applications.
  • Solid, easy-to-handle form for storage and weighing.
  • Compatible with solution-phase and solid-phase synthesis.
Catalog No. 50-002-83032