Learn More
Medchemexpress LLC Fmoc-Pen(Trt)-OH | 201531-88-6 | >99% | 613.76 | 1 G

Supplier: Medchemexpress LLC HYW0088761G
Fmoc-Pen(Trt)-OH is an amino acid derivative with an Fmoc protecting group that can be used to synthesize the inhibitory cystine knot (ICK) peptide ProTx-II.
- High purity.
- Suitable for peptide synthesis.
- Store at room temperature for 3 years.
- Soluble in DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.