Learn More
Medchemexpress LLC Fmoc-Phe(4-Br)-OH | 198561-04-5 | 99.83% | 466.32 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0137265G
Encompass_Preferred
Fmoc-Phe(4-Br)-OH is a widely used amino acid derivative. It can be utilized in the synthesis of compounds, for studying protein-protein interactions, and in the development of biomaterials. Additionally, it can be incorporated into peptide chains during peptide synthesis.
  • Used in the synthesis of compounds.
  • Applicable for the study of protein-protein interactions.
  • Aids in the development of biomaterials.
  • Can be incorporated into peptide chains during peptide synthesis.
Catalog No. 50-004-54468