Learn More
Medchemexpress LLC Fmoc-Phe-OH-13C | 916262-84-5 | 99.8% | C2313CH21NO4 | 50 MG

Supplier: Medchemexpress LLC HY79131S550MG
Fmoc-Phe-OH-13C is a 13C-labeled L-Phenylalanine derivative. This chemical is supplied as a white to off-white solid, with a high purity of 99.8% (HPLC) and 99% isotopic enrichment. It is suitable for various laboratory chemical applications and the manufacture of substances.
- High purity of 99.8% (HPLC)
- Isotopic enrichment of 99%
- White to off-white solid appearance
- Stable under recommended storage conditions
- Suitable for laboratory chemical applications
- Melting/freezing point: 183-184°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.