Learn More
Medchemexpress LLC Fmoc-Phe-OH-13C9,15N | 1217455-27-0 | 99.3% | C1513C9H2115NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY79131S15MG
Encompass_Preferred
Fmoc-Phe-OH-13C9,15N is a 15N-labeled and 13C-labeled Fmoc-Phe-OH. It is a phenylalanine derivative intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-35071