Learn More
Apexbio Technology LLC Fmoc-Phe-OH 35661-40-6 500g
SDP
Supplier:  Apexbio Technology LLC A7947500000
Encompass_Preferred
Fmoc-Phe-OH (CAS 35661-40-6) is a phenylalanine derivative used predominantly in solid-phase peptide synthesis It features an N-terminal fluorenylmethyloxycarbonyl (Fmoc) protecting group which allows selective deprotection under mild basic conditions This enables the controlled stepwise elongation of peptide chains through iterative deprotection-coupling cycles Due to its reliable protection strategy Fmoc-Phe-OH is widely applied in biochemical and pharmacological research for the synthesis of peptides peptide analogs and derivatives of biologically active molecules Its applications include drug discovery programs studies of receptor-ligand interactions and structure-function analyses of peptide-based biomolecules
Catalog No. 50-000-90161