Learn More
Apexbio Technology LLC Fmoc-Phe-Wang resin 25g
SDP
Supplier:  Apexbio Technology LLC A683125000
Encompass_Preferred
Fmoc-Phe-Wang resin is a polymeric solid-phase synthesis support utilized in peptide research It is used in solid-phase peptide synthesis (SPPS) for the stepwise construction of peptide chains by sequential coupling of amino acids after removal of the N-terminal Fmoc protecting group under alkaline conditions The anchored peptide can be released via mild acidic cleavage Based on these chemical properties Fmoc-Phe-Wang resin holds research potential in applications such as peptide-based therapeutic candidate development bioactive peptide screening and structural-functional analyses in protein research
Catalog No. 50-000-93383