Learn More
Medchemexpress LLC Fmoc-PNA-C(Bhoc)-OH | 186046-81-1 | MFCD22373799 | 95.6% | 701.72 | C39H35N5O8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW54012250MG
Encompass_Preferred
Fmoc-PNA-C(Bhoc)-OH is an Fmoc-protected peptide nucleic acid monomer with a Bhoc-protected cytosine base, provided as a white to off-white solid for research use. It is intended as a building block for the solid-phase synthesis of PNA oligomers and includes documented purity and storage information.
  • Fmoc-protected monomer for solid-phase synthesis.
  • Bhoc-protected cytosine nucleobase to prevent side reactions.
  • High purity suitable for oligonucleotide assembly.
  • Storage conditions provided for powder and in-solution forms.
  • Available in multiple milligram pack sizes for research workflows.
Catalog No. 50-002-84470