Learn More
Medchemexpress LLC Fmoc-pna-c(bhoc)-oh | 186046-81-1 | MFCD22373799 | 95.6% | 701.72 g·mol⁻¹ | C39H35N5O8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW540122100MG
Encompass_Preferred
Fmoc-PNA-C(Bhoc)-OH is a protected peptide nucleic acid (PNA) monomer designed for use as a building block in solid-phase PNA synthesis. The Fmoc protecting group allows standard Fmoc-based coupling and deprotection workflows, while the Bhoc protection on the nucleobase reduces side reactions during assembly. The monomer is intended for research applications, including construction of PNA oligomers for hybridization, antisense, and diagnostic probe development.
  • Fmoc-protected for compatibility with Fmoc solid-phase synthesis
  • Bhoc-protected nucleobase to minimize side reactions
  • Suitable for construction of PNA oligomers for hybridization and antisense studies
  • Stable monomer for reliable coupling efficiency
  • Supplied as a solid for convenient handling and storage
  • Intended for research use
Catalog No. 50-002-84205