Learn More
Medchemexpress LLC Fmoc-Pro-OH-13C5,15N | 1217452-48-6 | 343.33 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW013780S150MG
Encompass_Preferred
Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Pro-OH. Fmoc-Pro-OH is a proline derivative. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Used as tracers for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54450