Learn More
Medchemexpress LLC Fmoc-(R)-2-(pentenyl)Ala-OH | 288617-77-6 | MFCD09879889 | 98.8% | 379.45 | C23H25NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW018847100MG
Encompass_Preferred
Fmoc-(R)-2-(pentenyl)Ala-OH is an Fmoc-protected, R-configured alkenyl amino acid used as a peptide building block. It introduces a 4-pentenyl side chain for downstream modifications such as ring-closing metathesis and is supplied as a white to off-white solid with high HPLC purity.
  • Fmoc-protected for compatibility with standard Fmoc solid-phase peptide synthesis.
  • R-configuration supports stereospecific incorporation into peptides.
  • Alkenyl 4-pentenyl side chain enables stapled peptide synthesis via ring-closing metathesis.
  • High purity suitable for research synthesis (COA-reported HPLC purity).
  • Available in multiple small-scale pack sizes for peptide assembly workflows.
Catalog No. 50-003-01366