Learn More
STA PHARMACEUTICAL US LLC Fmoc-(R)-3-Amino-3-(2-fluoro-phenyl)-propionic acid | 25 g | CAS 511272-50-7 | MDL MFCD03428031
Supplier: STA PHARMACEUTICAL US LLC STAAA1117925GM
Fmoc-(R)-3-Amino-3-(2-fluoro-phenyl)-propionic acid is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 511272-50-7
- MDL: MFCD03428031
- InChIKey: LYKQNUSJPNQQLZ-JOCHJYFZSA-N
- Molecular Weight: 405.425
- Molecular Formula: C24H20FNO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid
- SMILES: O=C(C[C@@H](NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C4=CC=CC=C4F)O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.