Learn More
Apexbio Technology LLC Fmoc-Ser(HPO3Bzl)-OH 158171-14-3 10g
SDP
Supplier:  Apexbio Technology LLC A795910000
Encompass_Preferred
Fmoc-Ser HPO3Bzl -OH (CAS 158171-14-3) is a protected derivative of serine used in peptide chemistry It features an N-terminal Fmoc group and a benzyl-protected phosphate at the serine side chain allowing site-specific incorporation of phosphoserine into peptides during solid-phase synthesis Based on these properties Fmoc-Ser HPO3Bzl -OH holds research potential in studies of kinase-substrate interactions signal transduction protein phosphorylation states and the structural-functional analysis of phosphorylated proteins
Catalog No. 50-000-92558