Learn More
Medchemexpress LLC Fmoc-Ser(tBu)-OH-13C3,15N | 1217448-78-6 | 387.41 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW007941S10MG
Encompass_Preferred
Fmoc-Ser(tBu)-OH-13C3,15N is a 15N-labeled and 13C-labeled serine derivative, used as a laboratory chemical for the manufacture of substances.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of nitrogen and carbon.
  • Stable under recommended storage conditions.
  • For research use only.
Catalog No. 50-004-46534