Learn More
Apexbio Technology LLC Fmoc-Ser(tBu)-OH 71989-33-8 25g
SDP
Supplier:  Apexbio Technology LLC A796025000
Encompass_Preferred
Fmoc-Ser tBu -OH (71989-33-8) is a serine derivative modified with an N-terminal fluorenylmethoxycarbonyl (Fmoc) group and a tert-butyl (tBu) side-chain protecting group It is specifically designed for use in solid-phase peptide synthesis where the Fmoc group enables selective mild alkaline removal for stepwise peptide elongation and the tBu group allows for acid-mediated deprotection to expose the serine residue Based on these chemical properties Fmoc-Ser tBu -OH holds research potential in peptide and protein synthesis for applications such as protein structure determination interaction assays enzymatic activity assessments drug discovery bioassay development and studies of protein interactions and signal transduction pathways
Catalog No. 50-224-0411