Learn More
Apexbio Technology LLC Fmoc-Thr-OH 73731-37-0 25g
SDP
Supplier:  Apexbio Technology LLC A796725000
Encompass_Preferred
Fmoc-Thr-OH (CAS 73731-37-0) is a threonine derivative whose amino group is protected by the fluorenylmethyloxycarbonyl (Fmoc) group This protective group enables selective and controlled reactions during solid-phase peptide synthesis by temporarily masking the amino functionality Fmoc-Thr-OH is widely utilized in the construction of peptides with defined amino acid sequences Based on these properties Fmoc-Thr-OH facilitates structural and functional analyses of peptides supports peptide-based drug discovery and aids in the investigation of protein-ligand interactions in biomedical research contexts
Catalog No. 50-000-94165