Learn More
Medchemexpress LLC Fmoc-Thr(tBu)-OH | 71989-35-0 | 397.46 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007706
Encompass_Preferred
Fmoc-Thr(tBu)-OH is a threonine derivative used in custom peptide synthesis. It is also recognized as beneficial for ergogenic dietary substances. Amino acid derivatives, including Fmoc-Thr(tBu)-OH, are commercially utilized as ergogenic supplements because they influence the secretion of anabolic hormones, provide fuel during exercise, improve mental performance during stress-related tasks, and help prevent exercise-induced muscle damage.
  • Threonine derivative
  • Suitable for custom peptide synthesis
  • Functions as an ergogenic dietary substance
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Helps prevent exercise-induced muscle damage
Catalog No. 50-004-49865