Learn More
Medchemexpress LLC Fmoc-Tpi-OH | 204322-23-6 | 99.2% | 438.47 | 10 G

Supplier: Medchemexpress LLC HY13854610G
Fmoc-Tpi-OH is a biologically active amino acid designed for the synthesis of antimicrobial peptide mimetics.
- Biologically active amino acid
- Used to synthesize antimicrobial peptide mimetics
- White to off-white solid appearance
- High purity of 99.24% (HPLC)
- Recommended storage at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.